siRNA Details
Gene Symbol: CCDC107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195200.1 | NP_001182129.1 | 6 | 644 | ||
NM_001195201.1 | NP_001182130.1 | 6 | 624 | |||
NM_001195217.1 | NP_001182146.1 | 5 | 697 | |||
NM_174923.2 | NP_777583.2 | 5 | 725 | |||
XM_005251403.4 | XP_005251460.1 | 5 | 708 | |||
GenBank mRNA | AK075523.1 | - | 5 | 683 | ||
BC018758.1 | - | 5 | 701 | 4907976 | ||
BC051285.1 | - | 6 | 611 | 6586677 | ||
BE903559.1 | - | 6 | 633 | |||
DQ884403.1 | - | 6 | 554 |
