siRNA Details
Gene Symbol: GABPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002041.4 | NP_002032.2 | 8 | 1459 | ||
NM_016655.4 | NP_057739.1 | 8 | 1423 | |||
NM_181427.3 | NP_852092.1 | 9 | 1447 | |||
GenBank mRNA | BC016910.1 | - | 8 | 1144 | 4428422 | |
BT006652.1 | - | 7 | 1035 | |||
D13316.1 | - | 9 | 1137 | |||
JQ313901.1 | - | 7 | 996 | |||
U13047.1 | - | 8 | 1204 | |||
U13048.1 | - | 8 | 1168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
