siRNA Details
Gene Symbol: KLRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC093644.1 | - | 6 | 738 | 7939489 | |
BC112039.1 | - | 6 | 750 | 8327507 | ||
X54869.1 | - | 6 | 714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
