siRNA Details
Gene Symbol: BMP8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181809.3 | NP_861525.2 | 7 | 1621 | ||
GenBank mRNA | AY303954.1 | - | 7 | 1621 | ||
BC131617.1 | - | 7 | 937 | 40116584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
