siRNA Details
Gene Symbol: PRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261399.2 | NP_001248328.1 | 2 | 116 | ||
NM_002723.5 | NP_002714.2 | 2 | 116 | |||
GenBank mRNA | BC035362.1 | - | 2 | 102 | ||
BC128191.1 | - | 2 | 116 | |||
BC130386.1 | - | 2 | 101 | 40146417 | ||
K03207.1 | - | 2 | 115 |
