siRNA Details
Gene Symbol: NDUFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199987.1 | NP_001186916.1 | 2 | 339 | ||
NM_002493.4 | NP_002484.1 | 3 | 432 | |||
GenBank mRNA | AF035840.1 | - | 3 | 376 | ||
AF067167.1 | - | 3 | 405 | |||
AK289702.1 | - | 3 | 386 | |||
AW160372.1 | - | 2 | 270 | |||
BC009801.1 | - | 3 | 411 | 4334414 | ||
BC016708.1 | - | 3 | 446 | |||
BC029247.1 | - | 3 | 377 | 5171627 | ||
BP208846.1 | - | 3 | 430 | |||
CR456920.1 | - | 3 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
