siRNA Details
Gene Symbol: PPP1CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008709.1 | NP_001008709.1 | 2 | 296 | ||
NM_002708.3 | NP_002699.1 | 2 | 263 | |||
GenBank mRNA | AK098311.1 | - | 4 | 1072 | ||
AK127616.1 | - | 3 | 480 | |||
AK313586.1 | - | 2 | 172 | |||
AL557336.3 | - | 2 | 287 | |||
BC001888.1 | - | 2 | 184 | 3546588 | ||
BC004482.2 | - | 2 | 204 | 3689198 | ||
BC008010.1 | - | 2 | 204 | 3528742 | ||
BG480286.1 | - | 2 | 262 | |||
BT006629.1 | - | 2 | 140 | |||
J04759.1 | - | 1 | 74 | |||
M63960.1 | - | 2 | 169 | |||
S57501.1 | - | 2 | 184 | |||
X70848.1 | - | 2 | 167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
