siRNA Details
Gene Symbol: ULK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003565.2 | NP_003556.1 | 2 | 402 | ||
XM_011538798.2 | XP_011537100.1 | 2 | 179 | |||
XM_011538799.2 | XP_011537101.1 | 2 | 179 | |||
GenBank mRNA | AB018265.2 | - | 2 | 184 | ||
AF045458.1 | - | 2 | 402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
