siRNA Details
Gene Symbol: CCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002983.2 | NP_002974.1 | 3 | 722 | ||
GenBank mRNA | BC071834.1 | - | 3 | 694 | 5935287 | |
BX103455.1 | - | 3 | 722 | |||
D00044.1 | - | 3 | 703 | |||
M23452.1 | - | 3 | 703 | |||
M25315.1 | - | 3 | 703 | |||
X03754.1 | - | 3 | 703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
