siRNA Details
Gene Symbol: HIST1H2BI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003525.2 | NP_003516.1 | 1 | 386 | ||
GenBank mRNA | BC101653.1 | - | 1 | 446 | 8069159 | |
BC101655.1 | - | 1 | 446 | 8069161 |
