siRNA Details
Gene Symbol: HIST2H4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003548.2 | NP_003539.1 | 1 | 171 | ||
GenBank mRNA | BC019846.1 | - | 1 | 150 | ||
BC093969.1 | - | 1 | 165 | 7939814 | ||
BC108260.1 | - | 1 | 160 | |||
BC112193.1 | - | 1 | 165 | 8327661 | ||
CR542172.1 | - | 1 | 143 |
Gene Symbol: HIST2H4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034077.4 | NP_001029249.1 | 1 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
