siRNA Details
Gene Symbol: RFXANK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278727.1 | NP_001265656.1 | 9 | 1350 | ||
NM_001278728.1 | NP_001265657.1 | 8 | 1185 | |||
NM_003721.3 | NP_003712.1 | 10 | 1416 | |||
NM_134440.2 | NP_604389.1 | 9 | 1347 | |||
XM_005260134.4 | XP_005260191.1 | 10 | 1078 | |||
XM_005260135.3 | XP_005260192.1 | 10 | 1078 | |||
XM_005260136.4 | XP_005260193.1 | 10 | 1377 | |||
XM_005260137.4 | XP_005260194.1 | 10 | 1148 | |||
XM_006722930.3 | XP_006722993.1 | 9 | 1246 | |||
XM_017027415.1 | XP_016882904.1 | 9 | 1239 | |||
XM_017027416.1 | XP_016882905.1 | 8 | 1173 | |||
GenBank mRNA | AF077196.1 | - | 9 | 1064 | ||
AF094760.1 | - | 10 | 1328 | |||
AF105427.1 | - | 9 | 1073 | |||
AF105428.1 | - | 8 | 987 | |||
X75690.1 | - | 1 | 109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
