siRNA Details
Gene Symbol: S100A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC068064.1 | - | 2 | 436 | 6074799 | |
BC070291.1 | - | 4 | 677 | 6713909 | ||
X99920.1 | - | 3 | 454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
