siRNA Details
Gene Symbol: FOXD4L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199135.4 | NP_954586.4 | 1 | 163 | ||
GenBank mRNA | AY344641.1 | - | 1 | 163 | ||
AY344642.1 | - | 1 | 163 |
Gene Symbol: FOXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207305.4 | NP_997188.2 | 1 | 376 | ||
GenBank mRNA | AY344640.1 | - | 1 | 164 | ||
BC089432.1 | - | 1 | 379 | 30555309 | ||
BC103886.2 | - | 1 | 77 | |||
BC136570.1 | - | 1 | 111 | 9020560 | ||
BC136571.1 | - | 1 | 111 | 9020561 |
Gene Symbol: FOXD4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012184.4 | NP_036316.1 | 1 | 251 | ||
GenBank mRNA | AF452723.1 | - | 1 | 99 | ||
AK122950.1 | - | 1 | 251 | |||
AK125398.1 | - | 1 | 670 | |||
AY344639.1 | - | 1 | 163 | |||
BC136428.1 | - | 1 | 243 | 9020417 |
Gene Symbol: FOXD4L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199244.3 | NP_954714.2 | 1 | 78 |
Gene Symbol: FOXD4L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126334.1 | NP_001119806.1 | 1 | 910 |
Gene Symbol: FOXD4L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085476.1 | NP_001078945.1 | 1 | 670 | ||
GenBank mRNA | BC103887.2 | - | 1 | 78 | 40004959 | |
BC137365.1 | - | 1 | 158 | 9021363 | ||
BC137366.1 | - | 1 | 158 | 9021364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
