siRNA Details
Gene Symbol: LCE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178429.3 | NP_848516.1 | 1 | 173 | ||
GenBank mRNA | BC125197.1 | - | 1 | 173 |
Gene Symbol: LCE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014357.4 | NP_055172.1 | 2 | 192 | ||
GenBank mRNA | AF005080.1 | - | 2 | 189 | ||
AF086289.1 | - | 2 | 192 | |||
BC117235.1 | - | 2 | 183 | |||
BC117237.1 | - | 2 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
