siRNA Details
Gene Symbol: CTDNEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143775.1 | NP_001137247.1 | 8 | 1456 | ||
NM_015343.4 | NP_056158.2 | 9 | 1523 | |||
GenBank mRNA | AJ011916.1 | - | 8 | 1106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
