siRNA Details
Gene Symbol: STK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032296.3 | NP_001027467.2 | 2 | 424 | ||
NM_001286649.1 | NP_001273578.1 | 2 | 424 | |||
NM_003576.4 | NP_003567.2 | 2 | 368 | |||
XM_005254079.4 | XP_005254136.1 | 2 | 245 | |||
XM_017020794.1 | XP_016876283.1 | 2 | 541 | |||
XM_017020795.1 | XP_016876284.1 | 2 | 203 | |||
GenBank mRNA | AF024636.1 | - | 2 | 137 | ||
AF083420.1 | - | 2 | 240 | |||
AK297583.1 | - | 2 | 140 | |||
AK299140.1 | - | 2 | 141 | |||
BC035578.1 | - | 2 | 372 | 5539375 | ||
BT019619.1 | - | 2 | 59 | |||
CB996892.1 | - | 451 | ||||
CR542227.1 | - | 2 | 59 | |||
DA386652.1 | - | 2 | 330 | |||
DA395717.1 | - | 2 | 368 | |||
DB479387.1 | - | 2 | 417 | |||
EU668350.1 | - | 2 | 415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
