siRNA Details
Gene Symbol: FOXD4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012184.4 | NP_036316.1 | 1 | 328 | ||
GenBank mRNA | AF452723.1 | - | 1 | 176 | ||
AK122950.1 | - | 1 | 328 | |||
AK125398.1 | - | 1 | 747 | |||
AY344639.1 | - | 1 | 240 | |||
BC136428.1 | - | 1 | 320 | 9020417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
