siRNA Details
Gene Symbol: DHRS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005794.3 | NP_005785.1 | 7, 8 | 1128 | ||
XM_005267249.1 | XP_005267306.1 | 7, 8 | 840 | |||
XM_006720001.3 | XP_006720064.1 | 7, 8 | 803 | |||
XM_011536338.1 | XP_011534640.1 | 6, 7 | 769 | |||
XM_011536339.2 | XP_011534641.1 | 6, 7 | 619 | |||
GenBank mRNA | AB209653.1 | - | 5, 6 | 4770 | ||
AK092834.1 | - | 3, 4 | 2539 | |||
AK222857.1 | - | 7, 8 | 1128 | |||
AK309945.1 | - | 7, 8 | 2615 | |||
BC007339.2 | - | 6, 7 | 654 | |||
U31875.1 | - | 7, 8 | 1114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
