siRNA Details
Gene Symbol: DYRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004023.1 | NP_001004023.1 | 4 | 511 | ||
NM_003582.2 | NP_003573.2 | 3 | 417 | |||
XM_005273315.4 | XP_005273372.1 | 4 | 355 | |||
XM_011510061.2 | XP_011508363.1 | 3 | 251 | |||
GenBank mRNA | AF186773.1 | - | 3 | 394 | ||
AF186774.1 | - | 4 | 660 | |||
AF327561.1 | - | 4 | 289 | |||
AK291189.1 | - | 4 | 511 | |||
AK308082.1 | - | 3 | 417 | |||
AK313245.1 | - | 3 | 417 | |||
AU126055.1 | - | 3 | 417 | |||
AY590695.1 | - | 3 | 416 | |||
BC015501.1 | - | 4 | 468 | 3913541 | ||
Y12735.2 | - | 3 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
