siRNA Details
Gene Symbol: COA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040431.2 | NP_001035521.1 | 2 | 656 | ||
GenBank mRNA | AF070665.1 | - | 2 | 631 | ||
AK225611.1 | - | 2 | 635 | |||
AL524532.3 | - | 2 | 656 | |||
AU122748.1 | - | 2 | 622 | |||
BC002698.2 | - | 2 | 617 | 3609536 | ||
L38939.1 | - | 1 | 367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
