siRNA Details
Gene Symbol: ISCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000574.1 | - | 5 | 917 | ||
AY009127.1 | - | 6 | 1025 | |||
AY009128.1 | - | 5 | 929 | |||
BC011906.2 | - | 5 | 930 | 4136262 | ||
BG621612.1 | - | 543 | ||||
U47101.1 | - | 4 | 777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
