siRNA Details
Gene Symbol: OR3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002550.2 | NP_002541.2 | 1 | 718 | ||
XM_017024695.1 | XP_016880184.1 | 2 | 809 | |||
XM_017024696.1 | XP_016880185.1 | 2 | 783 | |||
GenBank mRNA | BC069414.1 | - | 1 | 718 | 7262242 | |
BC096187.3 | - | 1 | 718 | 40004009 | ||
BC096188.3 | - | 1 | 718 | 40004010 | ||
BC096189.3 | - | 1 | 718 | 40004011 | ||
BC096190.1 | - | 1 | 718 | 40004012 |
Gene Symbol: OR1M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004456.1 | NP_001004456.1 | 1 | 709 | ||
GenBank mRNA | BC137551.1 | - | 1 | 775 | 9021551 | |
BC137555.1 | - | 1 | 775 | 9021555 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146709, BC138440, BC138439.
Gene Ontology Categories:
