siRNA Details
Gene Symbol: MCUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017918.4 | NP_060388.2 | 3, 4 | 478 | ||
XM_006714246.3 | XP_006714309.1 | 3, 4 | 589 | |||
GenBank mRNA | AK000654.1 | - | 1, 2 | 165 | ||
AK095936.1 | - | 3, 4 | 385 | |||
AK291098.1 | - | 3, 4 | 472 | |||
AK311030.1 | - | 3, 4 | 342 | |||
BC002633.2 | - | 3, 4 | 416 | 3613298 | ||
CN427680.1 | - | 3, 4 | 482 | |||
CR457234.1 | - | 1, 2 | 75 |
