siRNA Details
Gene Symbol: PID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511384.2 | XP_011509686.1 | 1 | 158 | ||
GenBank mRNA | AK000708.1 | - | 1 | 114 | ||
AY317148.1 | - | 1 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
