siRNA Details
Gene Symbol: C19orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100418.1 | NP_001093888.1 | 4, 5 | 644 | ||
NM_001100419.1 | NP_001093889.1 | 4, 5 | 578 | |||
XM_005259977.1 | XP_005260034.1 | 2, 3 | 565 | |||
GenBank mRNA | AK000857.1 | - | 4, 5 | 532 | ||
BC012078.1 | - | 4, 5 | 533 | 4661993 | ||
BI836487.1 | - | 4, 5 | 455 | |||
KP143545.1 | - | 4, 5 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
