siRNA Details
Gene Symbol: DBNDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048225.2 | NP_001041690.2 | 1 | 450 | ||
NM_001048226.2 | NP_001041691.2 | 1 | 450 | |||
NM_018478.3 | NP_060948.3 | 2 | 496 | |||
GenBank mRNA | AF220191.1 | - | 2 | 502 | ||
AY113697.1 | - | 1 | 102 | |||
BI666711.1 | - | 1 | 461 | |||
BP308674.1 | - | 1 | 133 | |||
BQ787725.1 | - | 1 | 448 | |||
DT216122.1 | - | 1 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
