siRNA Details
Gene Symbol: DDX60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017631.5 | NP_060101.3 | 38 | 6061 | ||
GenBank mRNA | AK000042.1 | - | 10 | 1862 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
