siRNA Details
Gene Symbol: CTAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020994.4 | NP_066274.2 | 2 | 561 | ||
GenBank mRNA | AJ012835.1 | - | 2 | 552 | ||
AJ223040.1 | - | 2 | 561 | |||
BC002833.2 | - | 2 | 515 | 3638129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
