siRNA Details
Gene Symbol: ZNF253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021047.2 | NP_066385.2 | 4 | 1875 | ||
GenBank mRNA | AF038951.1 | - | 4 | 1112 | ||
AK074757.1 | - | 5 | 1786 | |||
AK074872.1 | - | 1 | 1525 | |||
BC014148.2 | - | 4 | 1875 | |||
BC065572.1 | - | 2 | 1650 | 6171141 | ||
BC125063.1 | - | 4 | 1854 | 40117279 | ||
BC125064.1 | - | 2 | 1631 | 40117289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
