siRNA Details
Gene Symbol: CABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019855.4 | NP_062829.1 | 6 | 1171 | ||
XM_017026992.1 | XP_016882481.1 | 8 | 1372 | |||
GenBank mRNA | AF169159.1 | - | 6 | 1164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
