siRNA Details
Gene Symbol: CKLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016326.3 | NP_057410.1 | 2 | 335 | ||
NM_016951.3 | NP_058647.1 | 4 | 590 | |||
NM_181640.2 | NP_857591.1 | 3 | 431 | |||
NM_181641.2 | NP_857592.1 | 3 | 494 | |||
GenBank mRNA | AF057306.1 | - | 4 | 568 | ||
AF096895.2 | - | 3 | 425 | |||
AF135380.2 | - | 4 | 584 | |||
AF135381.2 | - | 2 | 329 | |||
AF145216.2 | - | 3 | 488 | |||
AF151058.1 | - | 4 | 587 | |||
AK290307.1 | - | 3 | 405 | |||
AY358999.1 | - | 4 | 567 | |||
BC004380.1 | - | 4 | 573 | 3639550 | ||
BC091478.1 | - | 3 | 418 | 30333801 | ||
BU189908.1 | - | 3 | 391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
