siRNA Details
Gene Symbol: MCUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017918.4 | NP_060388.2 | 4, 5 | 596 | ||
XM_006714246.3 | XP_006714309.1 | 4, 5 | 707 | |||
GenBank mRNA | AK000654.1 | - | 2, 3 | 283 | ||
AK095936.1 | - | 4, 5 | 503 | |||
AK291098.1 | - | 4, 5 | 590 | |||
AK311030.1 | - | 4, 5 | 460 | |||
BC002633.2 | - | 4, 5 | 534 | 3613298 | ||
CR457234.1 | - | 2, 3 | 193 |
