siRNA Details
Gene Symbol: C19orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100418.1 | NP_001093888.1 | 873 | |||
NM_001100419.1 | NP_001093889.1 | 807 | ||||
GenBank mRNA | AK000857.1 | - | 761 | |||
BC012078.1 | - | 762 | 4661993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
