siRNA Details
Gene Symbol: ATAD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170535.1 | NP_001164006.1 | 16 | 2492 | ||
NM_001170536.1 | NP_001164007.1 | 16 | 2322 | |||
NM_018188.3 | NP_060658.3 | 16 | 2636 | |||
GenBank mRNA | AK001571.1 | - | 16 | 2475 | ||
AK056099.1 | - | 16 | 2322 | |||
BC011814.2 | - | 16 | 2458 | 3162989 | ||
BC063607.1 | - | 16 | 2450 | 4939339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
