siRNA Details
Gene Symbol: HOXC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018953.3 | NP_061826.1 | 2 | 596 | ||
NR_003084.2 | 2 | 630 | ||||
GenBank mRNA | BC034922.1 | - | 2 | 372 | ||
BC044635.1 | - | 2 | 372 | |||
BM786543.1 | - | 2 | 509 | |||
CN309383.1 | - | 2 | 172 |
