siRNA Details
Gene Symbol: CTAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020994.4 | NP_066274.2 | 990 | |||
NM_172377.4 | NP_758965.2 | 761 | ||||
GenBank mRNA | AJ223040.1 | - | 990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
