siRNA Details
Gene Symbol: KRTAP1-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030967.2 | NP_112229.1 | 1 | 288 | ||
GenBank mRNA | AB052868.1 | - | 2 | 150 | ||
AB052934.1 | - | 1 | 288 | |||
AJ406926.1 | - | 1 | 257 | |||
BC069321.1 | - | 1 | 274 | |||
BC069450.1 | - | 1 | 274 | |||
BC105069.1 | - | 1 | 263 | 8144072 | ||
BC105071.1 | - | 1 | 263 | 8144074 |
Gene Symbol: KRTAP1-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257305.1 | NP_001244234.1 | 1 | 103 |
