siRNA Details
Gene Symbol: ZNF205-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024166.1 | 3 | 1151 | |||
NR_024167.1 | 3 | 1146 | ||||
GenBank mRNA | BC001809.2 | - | 3 | 1151 | ||
BC001824.2 | - | 3 | 1151 | |||
BC020202.1 | - | 2 | 634 | |||
BC121099.1 | - | 3 | 773 | |||
BC121100.1 | - | 3 | 778 |
