siRNA Details
Gene Symbol: SMG1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135305.1 | 5 | 665 | |||
NR_135317.1 | 3 | 860 | ||||
GenBank mRNA | AB055226.1 | - | 5 | 1076 | ||
AK097897.1 | - | 3 | 371 | |||
AK304513.1 | - | 5 | 647 | |||
AK316536.1 | - | 5 | 961 |
Gene Symbol: SMG1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002453.5 | 4 | 801 | |||
GenBank mRNA | AK289905.1 | - | 4 | 800 | ||
BC063703.1 | - | 4 | 801 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
