siRNA Details
Gene Symbol: PPP2R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002718.4 | NP_002709.2 | 2 | 912 | ||
XM_006713686.3 | XP_006713749.1 | 2 | 1323 | |||
XM_011512956.2 | XP_011511258.1 | 2 | 1087 | |||
XM_017006787.1 | XP_016862276.1 | 2 | 912 | |||
GenBank mRNA | AK293012.1 | - | 2 | 892 | ||
AK314243.1 | - | 2 | 840 | |||
BC015034.1 | - | 2 | 881 | |||
BC029409.1 | - | 2 | 820 | |||
BC065531.1 | - | 2 | 859 | 6161260 | ||
L07590.1 | - | 2 | 799 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
