siRNA Details
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024127.1 | 4, 5 | 537 | |||
GenBank mRNA | AF277181.1 | - | 4, 5 | 325 | ||
AY277594.1 | - | 2, 3 | 105 | |||
BC022205.1 | - | 4, 5 | 1166 | |||
BC034611.1 | - | 2, 3 | 1388 | |||
BC068048.1 | - | 4, 5 | 537 | |||
BC107730.1 | - | 1, 2 | 929 | |||
KJ782215.1 | - | 5, 6 | 1666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
