siRNA Details
Gene Symbol: ZNF675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138330.2 | NP_612203.2 | 3 | 365 | ||
XM_017026442.1 | XP_016881931.1 | 3 | 370 | |||
XM_017026443.1 | XP_016881932.1 | 3 | 370 | |||
XM_017026444.1 | XP_016881933.1 | 3 | 362 | |||
GenBank mRNA | AK093669.1 | - | 3 | 352 | ||
AK307845.1 | - | 3 | 293 | |||
AY044432.1 | - | 3 | 290 | |||
BC032394.1 | - | 3 | 347 | |||
DA777369.1 | - | 3 | 384 |
