siRNA Details
Gene Symbol: RBMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002897.4 | NP_002888.1 | 14 | 3366 | ||
NM_016836.3 | NP_058520.1 | 14 | 3375 | |||
XM_005246737.1 | XP_005246794.1 | 15 | 3134 | |||
XM_005246738.2 | XP_005246795.1 | 15 | 3469 | |||
XM_005246739.2 | XP_005246796.1 | 15 | 3339 | |||
XM_005246740.1 | XP_005246797.1 | 15 | 3125 | |||
XM_005246741.3 | XP_005246798.1 | 15 | 3336 | |||
XM_017004626.1 | XP_016860115.1 | 14 | 3412 | |||
GenBank mRNA | AL117617.1 | - | 1 | 417 | ||
BC065192.1 | - | 1 | 636 | |||
BC069687.1 | - | 1 | 583 | |||
BC069706.1 | - | 1 | 582 | |||
BC069763.1 | - | 1 | 582 | |||
BC080620.1 | - | 1 | 611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
