siRNA Details
Gene Symbol: CHST5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024533.4 | NP_078809.2 | 3 | 3206 | ||
GenBank mRNA | AF246718.1 | - | 3 | 3207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
