siRNA Details
Gene Symbol: SMG1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135305.1 | 5 | 667 | |||
NR_135317.1 | 3 | 862 | ||||
GenBank mRNA | AB055226.1 | - | 5 | 1078 | ||
AK097897.1 | - | 3 | 373 | |||
AK304513.1 | - | 5 | 649 | |||
AK316536.1 | - | 5 | 963 |
Gene Symbol: SMG1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002453.5 | 4 | 803 | |||
GenBank mRNA | AK289905.1 | - | 4 | 802 | ||
BC063703.1 | - | 4 | 803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
