siRNA Details
Gene Symbol: HAGHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290137.1 | NP_001277066.1 | 7, 8 | 826 | ||
NM_001290139.1 | NP_001277068.1 | 7, 8 | 896 | |||
NM_001323635.1 | NP_001310564.1 | 7, 8 | 931 | |||
NM_032304.3 | NP_115680.1 | 6, 7 | 857 | |||
XM_011522712.1 | XP_011521014.1 | 6, 7 | 1088 | |||
XM_017023774.1 | XP_016879263.1 | 7, 8 | 721 | |||
GenBank mRNA | AK098340.1 | - | 2, 3 | 1485 | ||
AK293575.1 | - | 6, 7 | 857 | |||
BC004353.1 | - | 6, 7 | 832 | 3351990 | ||
BC015008.1 | - | 6, 7 | 785 | 4862387 | ||
BG821643.1 | - | 7, 8 | 643 | |||
BI333301.1 | - | 6, 7 | 694 | |||
BI834617.1 | - | 2, 3 | 162 | |||
BQ067068.1 | - | 2, 3 | 144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
