siRNA Details
Gene Symbol: RAB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164522.2 | NP_001157994.1 | 6 | 2886 | ||
NM_001304839.1 | NP_001291768.1 | 5 | 2760 | |||
NM_177403.5 | NP_796377.3 | 5 | 2960 | |||
GenBank mRNA | AK122657.1 | - | 6 | 2202 | ||
AK127140.1 | - | 6 | 2276 | |||
BC007382.1 | - | 1 | 492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
