siRNA Details
Gene Symbol: PMCHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003922.1 | 4 | 1693 | |||
GenBank mRNA | AY008413.1 | - | 4 | 1693 | ||
EU921431.1 | - | 5 | 405 | |||
EU921432.1 | - | 4 | 301 | |||
EU921433.1 | - | 5 | 395 | |||
EU921434.1 | - | 4 | 301 |
Gene Symbol: PMCHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU921435.1 | - | 4 | 293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
