siRNA Details
Gene Symbol: TMEM25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144037.1 | NP_001137509.1 | 9 | 1453 | ||
NM_001144038.1 | NP_001137510.1 | 8 | 1318 | |||
XM_017018431.1 | XP_016873920.1 | 8 | 1205 | |||
GenBank mRNA | AK027305.1 | - | 9 | 1342 | ||
AK075437.1 | - | 9 | 1217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
